Learn More
Abeba™ The Original 7209 Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
Brand: Abeba™ 7209/40
Additional Details : Weight : 0.78000kg
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- Black
- Slip resistance outsole sole
- Sanitized treated
- Heel strap non-foldable, detachable
- Breathable lining with Silver ions
- Adjustable instep area
- Heel cushion
Specifications
CE EN ISO 20347:2012 OB A, FO SRC/li | |
Leather | |
Slip-resistant | |
Animal fats, oil and fuel |
Black | |
40 | |
7209 |