Learn More
Abeba™ The Original 7209 Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
Brand:  Abeba™ 7209/41
Additional Details : Weight : 0.78000kg
 View more versions of this product
Product Code. 15622618
  • 65.96€ / 1 pair
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • Black
  • Slip resistance outsole sole
  • Sanitized treated
  • Heel strap non-foldable, detachable
  • Breathable lining with Silver ions
  • Adjustable instep area
  • Heel cushion
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC/li
Leather
Slip-resistant
Animal fats, oil and fuel
Black
41
7209