Learn More
Scientific Industries SI™ Recessed Platform
Product Code. 15547325 Shop All Ohaus Products
Product Code. 15547325 Supplier Scientific Industries SI™ Supplier No. 83041433
  • ONLINE EXCLUSIVE PRICE
    132.30€
    112.14€ / Pack of 1
    Save 20.16€
    15% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Accommodates Vortex-Genie 2 and Vortex-Genie Pulse Foam Inserts. Scientific Industries™ Recessed Platform can also be used with elastic bands and other items included in the Large Sample Set.
  • Can be used with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse
  • Use in conjunction with either Foam Inserts or Elastic Bands
  • Included in the Large Sample Set
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Vortex-Genie 2, Vortex-Genie Pulse, foam inserts, elastic bands