Learn More
Cole-Parmer™ Benchtop Heat Protector, Glass Ceramic
Product Code. 11897232
Change view
Click to view available options
Length (Metric):
150 mm
228 mm
300 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11897232 150 mm Each
11817242 300 mm Each
11807242 228 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11897232 Supplier Cole-Parmer™ Supplier No. 00273ZC
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Keep your benchtop safe from hot labware with this Glass Ceramic Benchtop Heat Protector.
  • Plate has corner feet to prevent heat contact with benchtop.
  • Glass ceramic surface resists stains and is easy to clean.
  • Temperature range is from -392°F to +1200°F (-236°C to +649°C).
Specifications
Product Type Benchtop Heat Protector
Length (Metric) 150 mm
Width (Metric) 150 mm
Dimensions (L x W) 150 x 150 mm
Quantity 1 Ea.