Learn More
Brady™ Thermal Transfer Printable Labels, B-427
Product Code. 11502945 Shop All Brady Products
Change view
Click to view available options
Size (Metric):
12.7 x 36.5 mm (W x H)
19.05 x 23.7 mm (W x H)
19.05 x 44.45 mm (W x H)
20.3 x 36.5 mm (W x H)
20.32 x 36.5 mm (W x H)
25.4 x 25.4 mm
25.4 x 50.8 mm (W x H)
25.4 x 57.15 mm (W x H)
25.4 x 95.25 mm (W x H)
38.1 x 19.05 mm (W x H)
48.26 x 80.95 mm (W x H)
50.8 x 9.53 mm (W x H)
55.88 x 12.7 mm (W x H)
57.15 x 50.08 mm (W x H)
63.5 x 57.15 mm (W x H)
Labels per Row:
1
2
4
Quantity:
10,000/Pk.
1000/Pk.
1500/Pk.
3000/Pk.
3500/Pk.
5000/Pk.
Unit Size:
1 roll
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size (Metric) Labels per Row Quantity Unit Size
11502945 38.1 x 19.05 mm (W x H) 1 3000/Pk. 1 roll
11849644 25.4 x 25.4 mm 2 5000/Pk. 1 roll
11552935 55.88 x 12.7 mm (W x H) 1 3000/Pk. 1 roll
12385043 25.4 x 50.8 mm (W x H) 1 3000/Pk. 1 roll
11593734 63.5 x 57.15 mm (W x H) 1 1000/Pk. 1 roll
11503744 50.8 x 9.53 mm (W x H) 1 1000/Pk. 1 roll
11533814 19.05 x 23.7 mm (W x H) 4 10,000/Pk. 1 roll
11518274 12.7 x 36.5 mm (W x H) 1 5000/Pk. 1 roll
11528274 19.05 x 44.45 mm (W x H) 1 3500/Pk. 1 roll
11538274 20.3 x 36.5 mm (W x H) 1 3500/Pk. 1 roll
11548274 25.4 x 57.15 mm (W x H) 1 3000/Pk. 1 roll
11558274 25.4 x 95.25 mm (W x H) 1 3000/Pk. 1 roll
11568274 48.26 x 80.95 mm (W x H) 1 1500/Pk. 1 roll
11578274 57.15 x 50.08 mm (W x H) 1 1500/Pk. 1 roll
11548204 19.05 x 44.45 mm (W x H) 4 10,000/Pk. 1 roll
11558204 20.32 x 36.5 mm (W x H) 4 10,000/Pk. 1 roll
11568204 25.4 x 57.15 mm (W x H) 2 3000/Pk. 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Product Code. 11502945 Supplier Brady™ Supplier No. 062367
  • ONLINE EXCLUSIVE PRICE
    319.00€
    289.01€ / 1 rolls
    Save 29.99€
    9% Off
View all promotions
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ideal for a variety of applications. Brady™ Thermal Transfer Printable Labels are made of self-laminating vinyl
  • Available in various sizes
Specifications
Certifications/Compliance UL Recognized
Description Thermal Transfer Printable Labels
Adhesive Type Acrylic
Color White, Translucent
Finish Matte
Format Roll
Material Vinyl, Self-laminating
Print Technology Thermal Transfer
Product Type Thermal Transfer Printer Label
For Use With (Application) Wire and cable marking
For Use With (Equipment) BBP81;i5100;i7100;PR Plus;IP Series;THT 76 mm Core Printers;i7500
Labels per Row 1
Min. Operating Temperature (Metric) -70°C
Model THT-135-427-3-AW
No. per Pack 3000 Label(s) / Roll
Quantity 3000/Pk.
Shape Rectangle
Size 38.10 mm (W) x 19.05 mm (H)
Size (Metric) 38.1 x 19.05 mm (W x H)
Temperature Range (English) -94°F to +158°F
Temperature Range (Metric) -70°C to +70°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Vertical Repeat (Metric) 22.23 mm
Dimensions (L x W) 19.05 x 38.10 mm
Width (English) 0.75 in.
Width (Metric) 38.1 mm
Width (English) Printable Area 38.10 mm
Height (English) 1.5 in.
Height (Metric) 38.1 mm
Height (Metric) Printable Area 19.05 mm
Alternate Part Number THT-135-427-3-AW