Learn More
Thermographic Measurements Thermax 10 Level Temperature Indicator
Product Code. 17806088
Change view
Click to view available options
Temperature Range (Metric):
188°C to 249°C
40°C to 71°C
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Unit Size
17806088 40°C to 71°C Pack of 10
17816088 188°C to 249°C Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17806088 Supplier Thermographic Measurements Supplier No. M10STHERGAD01P1
  • ONLINE EXCLUSIVE PRICE
    11.65€
    9.91€ / Pack of 10
    Save 1.74€
    14% Off
View all promotions
Estimated Shipment: 15-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ten level temperature Indicator Strips used to check maximum temperature.Single use, irreversible threshold indicator reports the highest temperature reached
  • Immediate Response
  • Permanent Record of Highest Temperature Achieved
  • Oil, Water, and Steam Resistant
  • Inexpensive and Easy to Use
  • Self-adhesive
Specifications
Certifications/Compliance ISO9001
Disposable Single-use
For Use With (Application) Laboratory Research, Medical services, Instrumentation, Electronics Industry, Automotive, Aerospace, Steel, Metallurgy, Plastics Industry, Food Industry
Height (English) 2 in.
Height (Metric) 51 mm
Humidity Range 40 to 60%
Length (Metric) 18 mm
Material Polyester Film
Description Thermax 10 Level Temperature indicator Range A
Max. Temperature (English) 160°F
Max. Temperature (Metric) 71°C
Min. Temperature (English) 36°F
Min. Temperature (Metric) 2°C
Temperature Range (English) 104°F to 160°F
Temperature Range (Metric) 40°C to 71°C
Calibrated Yes
For Use With (Equipment) Electrical Equipment, Pumps, Braking systems, Water Pumps, Aeronautical Equpment, Military Equipment
Accuracy ±1°C
Subdivision 40, 42, 44, 46, 49, 54, 60, 62, 65, 71°C
Display Visual Indicator
Width (Metric) 18 mm
Scale °C and °F