Learn More
Thermographic Measurements Thermax 5 Level Temperature Indicator
Product Code. 17808758
Click to view available options
Temperature Range (Metric):
37°C to 46°C
Unit Size:
Pack of 10
Product Code. 17808758
Brand: Thermographic Measurements M05STHERGAD02P1
Only null left
Add to basket
Edge
Add to basket
Five level temperature Indicator Strips used to check maximum temperature. Single use, irreversible threshold indicator reports the highest temperature reached.
  • Immediate Response
  • Permanent Record of Highest Temperature Achieved
  • Oil, Water, and Steam Resistant
  • Inexpensive and Easy to Use
  • Self-adhesive
Specifications
Certifications/Compliance ISO9001
Disposable Single-use
For Use With (Application) Laboratory Research, Medical services, Instrumentation, Electronics Industry, Automotive, Aerospace, Steel, Metallurgy, Plastics Industry, Food Industry
Height (English) 0.91 in.
Height (Metric) 51 mm
Humidity Range 40 to 60%
Length (Metric) 51 mm
Material Polyester Film
Length (English) 2 in.
Description Thermax 5 Level Temperature indicator Range A
Max. Temperature (English) 115°F
Max. Temperature (Metric) 46°C
Min. Temperature (English) 36°F
Min. Temperature (Metric) 2°C
Temperature Range (English) 99°F to 115°F
Temperature Range (Metric) 37°C to 46°C
Calibrated Yes
For Use With (Equipment) Electrical Equipment, Pumps, Braking systems, Water Pumps, Aeronautical Equpment, Military Equipment
Accuracy ±1°C
Subdivision 37, 40, 42, 44, 46°C, 99, 104, 108, 111, 115°F
Display Visual Indicator
Width (Metric) 51 mm
Scale °C and °F