Learn More
Thermo Scientific™ BetaSil™ C18 Guard Cartridges
Product Code. 10427334 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric) Inner:
2.1 mm
3 mm
4 or 4.6 mm
Particle Size:
3 μm
5 μm
Unit Size:
Pack of 4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Particle Size unitSize
10427334 2.1 mm 3 μm Pack of 4
10621206 3 mm 5 μm Pack of 4
10448094 4 or 4.6 mm 5 μm Pack of 4
10035513 2.1 mm 5 μm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10427334 Supplier Thermo Scientific™ Supplier No. 70103012101
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Economically extends HPLC column lifetime
Features of Betasil C18 Solid Phase
  • Thermo Scientific Chemistry-on 100Å silica
  • High surface area with high bonded phase coverage
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 3 μm
Length (Metric) 10 mm
Diameter (Metric) Inner 2.1 mm
Stationary Phase C18
Product Line BetaSil