Learn More
Unity Lab Services™ Thermo Scientific™ Centrifugation Validation Service
Product Code. 15536209
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific Centrifugation
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15536209 Thermo Scientific Centrifugation Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15536209 Supplier Unity Lab Services™ Supplier No. IOQPCKE89003492
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 16-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Stay compliant with our Industry-Exclusive, No-Charge Requalification Guarantee included with each Thermo Scientific Centrifugation Validation Service for Benchtop, Micro, Bioprocessing, Clinical, and Standard floor model centrifuges: Thermo Scientific, Sorvall
IQ
  • Unpacking, Assembly and Installation Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order. Verifies equipment is assembled and installed and any initial diagnostics tests have been performed. Any abnormal event(s) observed during assembly and installation have been documented.
  • Utility, Facility and Environment Verifies that the installation site satisfactorily meets manufacturer-specified environmental requirements.
OQ
  • System Component Information Detailed configuration information for each system component have been recorded.
  • OQ Limits: Record a list of manufacturer recommended limits for ensuring that the system is operating as expected.
  • Equipment Operational Tests and Results: Test important equipment functions to verify that the equipment operates as intended by the manufacturer and required by the user. This is a group of selected important equipment parameters for testing according to the intended use of the equipment.
Key Tests: Min/Max Speed, Brief/Extended Time, Temperature for Refrigerated Models
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific Centrifugation
Product Type IQ/OQ Service