Learn More
Thermo Scientific™ Thermo Scientific™ Watch Glass
Product Code. 17145321
Change view
Click to view available options
Material:
Glass
Unit Size:
Pack of 400
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
17145321 Glass Pack of 400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17145321 Supplier Thermo Scientific™ Supplier No. WATCH GLASS
  • ONLINE EXCLUSIVE PRICE
    255.00€
    231.00€ / Pack of 400
    Save 24.00€
    9% Off
Estimated Shipment: 24-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Thermo Scientific™ Watch Glass is used as a beaker cover and used in weighing, crystallization, and drying in the lab.
Thermo Scientific™ Watch Glass is used as a beaker cover as well as used in weighing, crystallization, and drying in the lab.
  • WATCH GLASS 2 PER SET
  • 200 SETS /CS
TRUSTED_SUSTAINABILITY
Specifications
Description WATCH GLASS 2 PER SET, 200 SETS /CS
Material Glass
Type Watch Glass
For Use With (Application) Used as Beaker Covers