Learn More
Dostmann Electronic™ Digital Thermometer
Product Code. 11814372
Change view
Click to view available options
Calibrated:
Yes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Calibrated Unit Size
11814372 Yes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11814372 Supplier Dostmann Electronic™ Supplier No. 50000700
  • ONLINE EXCLUSIVE PRICE
    475.26€
    260.00€ / Pack of 1
    Save 215.26€
    45% Off
View all promotions
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
With temperature measurements over a range of -200°C to +850°C, this digital thermometer is ideal for use in many laboratory, scientific, and other research environments.
  • The all-round talent for temperature measurements with Pt100 sensor over a range of -200°C to +850°C
  • The thermocouple (type J, K, L, N, R, S, T) to +1760°C with a resolution of 0,1°C over the full measuring range
  • Ideal for quality assurance service and production
  • Integrated calibration function for simple compensation of sensor tolerances
  • Physical 1-point, 2-point or 3-point calibration function
Specifications
Calibrated Yes