Learn More
VEE GEE Thermometer Case, Nickel-plated Brass Armor
Shop All Thermo Scientific Incubateurs à CO2 ProductsDescription
- Armor features a threaded screw cap with ring top for suspension where necessary
- Thermometer Length: Option of 12 in (305 mm) or 15.5 in (393 mm)
- Outside Diameter: 0.374 in (9.5 mm / Inside Diameter: 0.295 in (7.5 mm)
Specifications
Specifications
| Disposable | Reusable |
| For Use With (Application) | Protect liquid-in-glass thermometers from rough handling and shock during use |
| Length (English) Overall | 12 in. |
| Length (Metric) | 30.5 cm |
| Material | Nickel-plated |
| Weight (Metric) | 70 g |
| Product Type | Thermometer Armor Case |
| For Use With (Equipment) | Liquid-in-glass thermometers |
| Width (Metric) | 1.9 cm |