Learn More
Sonoco™ ThermoSafe PolarPack™ Standard
Product Code. 13089332
Change view
Click to view available options
Dimensions (LxWxH):
10.5 x 7.5 x 1.25 in.
5.5 x 3.5 x 0.75 in.
6 x 5.5 x 1 in.
6.5 x 5.5 x 1 in.
8 x 5.5 x 1.25 in.
Weight (English):
6 oz.
12 oz.
16 oz.
20 oz.
48 oz.
Material:
Polyethylene
Polyethylene Film
Unit Size:
Pack of 12
Pack of 24
Pack of 36
Pack of 48
Pack of 768
Pack of 96
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (LxWxH) Weight (English) Material Unit Size
13089332 6 x 5.5 x 1 in. 12 oz. Polyethylene Pack of 48
12201382 6 x 5.5 x 1 in. 12 oz. Polyethylene Pack of 48
13099332 6.5 x 5.5 x 1 in. 16 oz. Polyethylene Film Pack of 36
17663585 8 x 5.5 x 1.25 in. 20 oz. Polyethylene Pack of 24
16383356 8 x 5.5 x 1.25 in. 20 oz. Polyethylene Pack of 768
13029342 10.5 x 7.5 x 1.25 in. 48 oz. Polyethylene Film Pack of 12
12201362 5.5 x 3.5 x 0.75 in. 6 oz. Polyethylene Film Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 13089332 Supplier Sonoco™ Supplier No. PP12
  • ONLINE EXCLUSIVE PRICE
    61.38€
    25.31€ / Pack of 48
    Save 36.07€
    58% Off
Estimated Shipment: 01-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
PolarPack refrigerants provide thermal solutions for temperature critical packages during transit. Available in a broad selection of styles and a multitude of sizes, our refrigerants are designed to provide reliable temperature protection for a vast array of temperature-sensitive products and packages.
  • Thermal solutions for temperature critical packages during transit
  • Gel will phase change at 0° C.
  • condition coolant in freezer before shipments.
  • refrigeration for transport of food, pharmaceuticals and medical products. 
  • For frozen or refrigerated shipments.
  • Alternative to wet ice.
  • Available in several sizes
TRUSTED_SUSTAINABILITY
Specifications
Weight (English) 12 oz.
Dimensions (LxWxH) 6 x 5.5 x 1 in.
Material Polyethylene
Freezing Point 0°C
For Use With (Application) For shipping temperature critical packages during transit