Learn More
SI Analytics™ Thermostatic Bath
Product Code. 11964709 Shop All SI Analytics Products
Change view
Click to view available options
For Use With (Application):
Opaque or translucent fluid
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
11964709 Opaque or translucent fluid Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11964709 Supplier SI Analytics™ Supplier No. 285418526
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
SI Analytics™ Thermostatic Bath features an acrylic glass container that can hold up to two measuring points.
  • Glass Panelled Thermostatic Bath
  • Working temperature 10°C to 60°C
  • Acrylic glass container for up to two measuring points
  • Heater with Pt 100-PID control
TRUSTED_SUSTAINABILITY
Specifications
Type Immersion Thermostat
For Use With (Application) Opaque or translucent fluid
Model CT 72/P
Temperature (Metric) Maximum 60°C
Electrical Requirements 230 V
Voltage 230 V