missing translation for 'onlineSavingsMsg'
Learn More

Thiamine Pyrophosphate Chloride, TRC

Product Code. 17954895
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17954895 1 g 1g
30010912 5 g 5g
17973905 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17954895 Supplier Toronto Research Chemicals Supplier No. TRCT3440701G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154-87-0
Molecular Formula C12 H19 N4 O7 P2 S . Cl
Molecular Weight (g/mol) 460.77
Synonym Thiazolium, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-(4,6,6-trihydroxy-4,6-dioxido-3,5-dioxa-4,6-diphosphahex-1-yl)-, chloride (1:1), Thiamine, trihydrogen pyrophosphate (ester) (8CI), Thiazolium, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-(4,6,6-trihydroxy-3,5-dioxa-4,6-diphosphahex-1-yl)-, chloride, P,P'-dioxide, Thiazolium, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-(4,6,6-trihydroxy-4,6-dioxido-3,5-dioxa-4,6-diphosphahex-1-yl)-, chloride (9CI), B-Neurox, Berolase, Biosyth, Bioxilasi, Bivitasi, Co-B1, Cocarbina, Cocarboxil, Cocarboxylase, Coenbione, Coxylase, Diphosphothiamin, Diphosphothiamine, Farmaneurina, Metabolase, Nutrase, Pyrolase, Pyruvodehydrase, TDP, TDP (thiamin ester), TPP, TPP (coenzyme), Thiamin pyrophosphate, Thiamine diphosphate, Thiamine diphosphate ester chloride, Thiamine diphosphoric acid ester chloride, Thiamine pyrophosphate, Thiamine pyrophosphate chloride, Thiamine pyrophosphate hydrochloride, Thiamine pyrophosphoric acid ester chloride, Thiamine pyrophosphoric ester, Thiamine-PP, Vitamin B1 pyrophosphate chloride
IUPAC Name 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethyl phosphono hydrogen phosphate;chloride
SMILES [Cl-].Cc1ncc(C[n+]2csc(CCOP(=O)(O)OP(=O)(O)O)c2C)c(N)n1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H18N4O7P2S.ClH/c1-8-11(3-4-22-25(20,21)23-24(17,18)19)26-7-16(8)6-10-5-14-9(2)15-12(10)13;/h5,7H,3-4,6H2,1-2H3,(H4-,13,14,15,17,18,19,20,21);1H
Formula Weight 460.0138
Percent Purity >95%
Chemical Name or Material Thiamine pyrophosphate chloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.