missing translation for 'onlineSavingsMsg'
Learn More

Thioacetic Acid, TRC

Product Code. 30455307
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455307 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455307 Supplier Toronto Research Chemicals Supplier No. TRCT343895100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ethanethioic acid
CAS 507-09-5
Molecular Formula C2 H4 O S
Quantity 100 g
Synonym Ethanethioic acid, Acetyl mercaptan, Ethanethiolic acid, Methanecarbothiolic acid, Monothioacetic acid, Thiacetic acid, Thioacetic acid, Thiolacetic acid, Thionoacetic acid
InChI Formula InChI=1S/C2H4OS/c1-2(3)4/h1H3,(H,3,4)
SMILES CC(=O)S
IUPAC Name ethanethioic S-acid
Molecular Weight (g/mol) 76.12
Formula Weight 75.9983
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.