missing translation for 'onlineSavingsMsg'
Learn More

Thiocolchicine, TRC

Product Code. 30451948
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451948 250 mg 250mg
30450352 10 mg 10mg
30449167 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30451948 Supplier Toronto Research Chemicals Supplier No. TRCT344390250MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2730-71-4
Molecular Formula C22 H25 N O5 S
Molecular Weight (g/mol) 415.5
Synonym Acetamide, N-[(7S)-5,6,7,9-tetrahydro-1,2,3-trimethoxy-10-(methylthio)-9-oxobenzo[a]heptalen-7-yl]-, Acetamide, N-[5,6,7,9-tetrahydro-1,2,3-trimethoxy-10-(methylthio)-9-oxobenzo[a]heptalen-7-yl]-, (S)-, Colchicine, 10-thio- (8CI), Colchicine, thio- (6CI), Benzo[a]heptalene, acetamide deriv., N-[(7S)-5,6,7,9-Tetrahydro-1,2,3-trimethoxy-10-(methylthio)-9-oxobenzo[a]heptalen-7-yl]acetamide, 10-Demethoxy-10-(methylthio)colchicine, NSC 186301, Thiocolchicine
IUPAC Name N-[(7S)-1,2,3-trimethoxy-10-methylsulfanyl-9-oxo-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide
SMILES COc1cc2CC[C@H](NC(=O)C)C3=CC(=O)C(=CC=C3c2c(OC)c1OC)SC

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C22H25NO5S/c1-12(24)23-16-8-6-13-10-18(26-2)21(27-3)22(28-4)20(13)14-7-9-19(29-5)17(25)11-15(14)16/h7,9-11,16H,6,8H2,1-5H3,(H,23,24)/t16-/m0/s1
Formula Weight 415.1453
Chemical Name or Material Thiocolchicine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.