missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Thioflavin T

Product Code. 30040179
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040179 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30040179 Supplier Tocris Bioscience™ Supplier No. 7122/10
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Cell-permeable fluorescent amyloid stain. Used for: in vitro Aβ staining in brain tissues.

Key information: Thioflavin T is a cell-permeable fluorescent amyloid stain.

Used for: staining of insoluble senile plaques of Aβ in brain tissues, monitoring the kinetics of in vitro polyglutamine amyloid formation of tNhtt-42Q aggregates in a Huntington's disease cell model. Thioflavin T is used to confirm formation of β sheet structure from mutant huntingtin exon-1 aggregates (Ex1Q48) in vitro. Application: fluorescence microscopy.

Properties and Photophysical Data: Thioflavin T is a cationic benzothiazole. It increases in fluorescence upon strong binding (Ki = 580 nM) to the stacked β sheets of amyloid fibrils. Excitation and emission maxima (λ) are 440 nm and 485 nm, respectively; quantum yield = 0.43; extinction coefficient = 31600 M-1cm-1.

This product is supplied with a high degree of hydration and some residual inorganic salts, the amount of which are batch dependent. Please refer to the Certificate of Analysis to obtain the batch specific Net Product Content.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[4-(Dimethylamino)phenyl]-3,6-dimethylbenzothiazolium chloride
CAS 2390-54-7
Quantity 10 mg
Target Fluorescent Amyloid B Probes
Molecular Formula C17H19ClN2S
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.