missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thionazin, TRC
Specifications
Specifications
| Chemical Name or Material | Thionazin |
| CAS | 297-97-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C8 H13 N2 O3 P S |
| Quantity | 250 mg |
| Synonym | Phosphorothioic acid','O,O-diethyl O-2-pyrazinyl ester, Ethyl pyrazinyl phosphorothioate (6CI), Phosphorothioic acid','O,O-diethyl O-pyrazinyl ester (8CI,9CI), AC 18133, ACC 18133, American Cyanamid 18133, CL 18133, Cynem, Cynophos, EN 18133, ENT 25,580, Experimental nematocide 18,133, Nemafos, Nemafos 10 G, Nemaphos, Nematocide GR, O,O-Diethyl O-2-pyrazinyl phosphorothioate, Thionazin, Thionazine, Zinophos, Zynophos |
| InChI Formula | InChI=1S/C8H13N2O3PS/c1-3-11-14(15,12-4-2)13-8-7-9-5-6-10-8/h5-7H,3-4H2,1-2H3 |
| SMILES | CCOP(=S)(OCC)Oc1cnccn1 |
| IUPAC Name | diethoxy-pyrazin-2-yloxy-sulfanylidene-λ^{5}-phosphane |
| Molecular Weight (g/mol) | 248.24 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?