missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiophanate-methyl-d6 (O,O-dimethyl-d6), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Thiophanate-methyl-d6 |
| CAS | 1398065-77-4 |
| Molecular Formula | C12 H8 D6 N4 O4 S2 |
| Quantity | 0.05 g |
| Synonym | N,N'-[1,2-Phenylenebis(iminocarbonothioyl)]biscarbamic-d6 Acid C,C'-Dimethyl Ester, 4,4'-o-Phenylenebis[3-thioallophanic-d6 Acid Dimethyl Ester, 1,2-Bis[3-(methoxycarbonyl)-2-thioureido]benzene-d6, 3336 G-d6, Caligran-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6, Topsin M 70 WP-d6 |
| InChI Formula | InChI=1 S/C12H14N4O4S2/c1-19-11(17)15-9(21)13-7-5-3-4-6-8(7)14-10(22)16-12(18)20-2/h3-6 H,1-2H3,(H2,13,15,17,21)(H2,14,16,18,22)/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])OC(=O)NC(=S)Nc1ccccc1NC(=S)NC(=O)OC([2 H])([2 H])[2 H] |
| IUPAC Name | trideuteriomethyl N-[[2-(trideuteriomethoxycarbonylcarbamothioylamino)phenyl]carbamothioyl]carbamate |
| Molecular Weight (g/mol) | 348.43 |
| Formula Weight | 348.0833 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction