missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-3-boronic acid, 98%

Product Code. 11410038
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410038 1 g 1g
11420038 5 g 5g
11430038 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410038 Supplier Thermo Scientific Alfa Aesar Supplier No. B23637.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6165-69-1 | C4H5BO2S | 127.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6165-69-1
Molecular Formula C4H5BO2S
Molecular Weight (g/mol) 127.95
MDL Number MFCD00151851
InChI Key QNMBSXGYAQZCTN-UHFFFAOYSA-N
Synonym 3-thiopheneboronic acid, thiophene-3-boronic acid, 3-thienylboronic acid, thiophene 3-boronic acid, thiophene-3-boronicacid, thiophen-3-yl-3-boronic acid, 3-thienyl boronic acid, 3-thiophene boronic acid, thiophen-3-yl-boranediol, boronic acid, 3-thienyl
PubChem CID 581760
IUPAC Name thiophen-3-ylboronic acid
SMILES OB(O)C1=CSC=C1

Specifications

Melting Point 164°C to 169°C
Quantity 1 g
Beilstein 113573
Formula Weight 127.96
Percent Purity 98%
Chemical Name or Material Thiophene-3-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.