missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioproperazine Dimesilate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Thioproperazine Dimesilate |
| CAS | 2347-80-0 |
| Analyte or Component Names | Thioproperazine Dimesilate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H30 N4 O2 S2 . 2 C H4 O3 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | 10H-Phenothiazine-2-sulfonamide, N,N-dimethyl-10-[3-(4-methyl-1-piperazinyl)propyl]-, methanesulfonate (1:2), 10H-Phenothiazine-2-sulfonamide, N,N-dimethyl-10-[3-(4-methyl-1-piperazinyl)propyl]-, dimethanesulfonate (9CI), Phenothiazine-2-sulfonamide, N,N-dimethyl-10-[3-(4-methyl-1-piperazinyl)propyl]-, dimethanesulfonate (6CI,7CI,8CI), Mageptyl, Majeptil, Thioperazine dimethanesulfonate, Thioproperazine bismethanesulfonate, Thioproperazine dimesylate, Thioproperazine dimethanesulfonate, Thioproperazine mesylate, Thioproperazine methanesulfonate, Vontil |
| InChI Formula | InChI=1S/C22H30N4O2S2.2CH4O3S/c1-23(2)30(27,28)18-9-10-22-20(17-18)26(19-7-4-5-8-21(19)29-22)12-6-11-25-15-13-24(3)14-16-25;2*1-5(2,3)4/h4-5,7-10,17H,6,11-16H2,1-3H3;2*1H3,(H,2,3,4) |
| Solution Type | Neat |
| SMILES | CN(C)S(=O)(=O)c1ccc2Sc3ccccc3N(CCCN4CCN(C)CC4)c2c1.CS(=O)(=O)O.CS(=O)(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?