missing translation for 'onlineSavingsMsg'
Learn More

Thioridazine Hydrochloride, TRC

Product Code. 30450455
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450455 10 g -
30449177 50 mg 50mg
30450696 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30450455 Supplier Toronto Research Chemicals Supplier No. TRCT36880010G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130-61-0
Molecular Formula C21 H26 N2 S2 . Cl H
Molecular Weight (g/mol) 407.04
Synonym Thioridazine Hydrochloride, 10H-Phenothiazine, 10-[2-(1-methyl-2-piperidinyl)ethyl]-2-(methylthio)-, monohydrochloride (9CI), Phenothiazine, 10-[2-(1-methyl-2-piperidyl)ethyl]-2-(methylthio)-, hydrochloride (6CI,7CI), Phenothiazine, 10-[2-(1-methyl-2-piperidyl)ethyl]-2-(methylthio)-, monohydrochloride (8CI), 10-[2-(1-Methylpiperidin-2-yl)ethyl]-2-(methylsulfanyl)-10H-phenothiazine hydrochloride, 10-[3-(1-Methyl-2-piperidyl)ethyl]-2-(methylthio)phenothiazine hydrochloride, 2-Methylmercapto-10-[2-(N-methyl-2-piperidyl)ethyl]phenothiazine hydrochloride, Aldazine, Mallorol, Mellaril, Mellaril hydrochloride, Melleretten, Melleril, Melleril (tablet), NSC 186060, Novoridazine, Orsanil, Ridazin, Stalleril, Thioridazine hydrochloride, Thioril, Thiozine, 10H-Phenothiazine, 10-[2-(1-methyl-2-piperidinyl)ethyl]-2-(methylthio)-, hydrochloride (1:1)
IUPAC Name 10-[2-(1-methylpiperidin-2-yl)ethyl]-2-methylsulfanylphenothiazine;hydrochloride
SMILES Cl.CSc1ccc2Sc3ccccc3N(CCC4CCCCN4C)c2c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C21H26N2S2.ClH/c1-22-13-6-5-7-16(22)12-14-23-18-8-3-4-9-20(18)25-21-11-10-17(24-2)15-19(21)23;/h3-4,8-11,15-16H,5-7,12-14H2,1-2H3;1H
Formula Weight 406.1304
Chemical Name or Material Thioridazine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.