missing translation for 'onlineSavingsMsg'
Learn More

Thiotriazinone, 98+%, Thermo Scientific Chemicals

Product Code. 11435287
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435287 10 g 10g
11445287 50 g 50g
11455287 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435287 Supplier Thermo Scientific Alfa Aesar Supplier No. B22663.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58909-39-0
Molecular Formula C4H5N3O2S
Molecular Weight (g/mol) 159.163
MDL Number MFCD00185756
InChI Key UMWWHOXOVPIGFD-UHFFFAOYSA-N
Synonym thiotriazinone, tetrahydro-2-methyl-3-thioxo-1,2,4-triazine-5,6-dione, unii-679wf9q9w1, 2-methyl-3-thioxo-1,2,4-triazinane-5,6-dione, 1,2,4-triazin-2-methyl-6-hydroxy-3-thio-5-one, 2-methyl-3-sulfanyl-1,2-dihydro-1,2,4-triazine-5,6-dione, 1,2,4-triazine-5,6-dione, tetrahydro-2-methyl-3-thioxo, acmc-20ajjm, ceftriaxone impurity c, ksc269g8j
PubChem CID 3034640
IUPAC Name 2-methyl-3-sulfanylidene-1,2,4-triazinane-5,6-dione
SMILES CN1C(=S)NC(=O)C(=O)N1

Specifications

Melting Point 168°C to 171°C
Quantity 10 g
Formula Weight 159.16
Percent Purity ≥98%
Chemical Name or Material Thiotriazinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.