Learn More
THOC5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565514
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: SDAEEEQTTKR RRPTLGVQLD DKRKEMLKRH PLSVMLDLKC KDDSVLHLTF YYLMNLNIMT VKAKVTTAME LITPISAGDL LSPDSVLSCL YPGDHGKKTP NPANQYQFDK VGILTLSDYV LELGHPYLWV QKLGG Highest antigen sequence identity to the following orthologs - mouse 97%, rat 96%.
Component the THO subcomplex of the TREX complex. The TREX complex specifically associates with spliced mRNA and not with unspliced pre-mRNA. It is recruited to spliced mRNAs by a transcription-independent mechanism. Binds to mRNA upstream of the exon-junction complex (EJC) and is recruited in a splicing- and cap-dependent manner to a region near the 5' end of the mRNA where it functions in mRNA export. The recruitment occurs via an interaction between THOC4 and the cap-binding protein NCBP1. The TREX complex is essential for the export of Kaposi's sarcoma-associated herpesvirus (KSHV) intronless mRNAs and infectious virus production. The recruitment of the TREX complex to the intronless viral mRNA occurs via an interaction beteen KSHV ORF57 protein and THOC4. UAP56 functions as a bridge between THOC4 and the THO complex. THOC5 in conjunction with THOC4 functions in NXF1-NXT1 mediated nuclear export of HSP70 mRNA. May be involved in the differentiation of granulocytes and adipocytes.Specifications
THOC5 | |
Polyclonal | |
Unconjugated | |
THOC5 | |
1700060C24Rik; A430085L24Rik; C22orf19; Fmip; Fms interacting protein; Fms-interacting protein; fSAP79; Functional spliceosome-associated protein 79; hTREX90; KIAA0983; NF2/meningioma region protein pK1.3; PK1.3; placental protein 39.2; PP39.2; serum/glucocorticoid regulated kinase 2; Sgk2; THO complex 5; THO complex subunit 5 homolog; Thoc5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8563 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13769 | |
THOC5 | |
Recombinant Human THOC5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only