Learn More
THSD7A Antibody (CL3779) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61639DL594
Product Code. 30295576
  • 737.00€ / 0.10mL
Description
THSD7A Monoclonal antibody specifically detects THSD7A in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
THSD7A
Monoclonal
DyLight 594
thrombospondin, type I, domain containing 7A
This antibody was developed using a recombinant protein derived from Q9UPZ6, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology
221981
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL3779
50mM Sodium Borate
Mouse
Protein A purified