missing translation for 'onlineSavingsMsg'
Learn More

Thymidylate Synthase Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Provide accurate, reproducible results Epredia™ Thymidylate Synthase Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-471-P1

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12681537

  • 484.00€ / 500µL
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Thymidylate Synthase (TS) (EC 2.1.1.45) converts deoxyuridine monophosphate (dUMP) to deoxythymidine monophosphate (dTMP), which is essential for DNA biosynthesis. TS is a critical target for the fluoropyrimidines, an important group of antineoplastic drugs that are widely used in the treatment of solid tumors. Both 5-FU and fluorodeoxyuridine are converted in tumor cells to FdUMP which inactivates TS by formation of a ternary covalent complex in the presence of the folate cofactor 5,10-methylenetetrahydrofolate. Expression of TS protein is associated with response to 5-fluorouracil (5-FU) in human colorectal, gastric, head and neck, and breast carcinomas.

Host Species: Mouse

Clone: TS 106

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human TS enzyme

Molecular Weight: 36kDa

Positive Control: 5-FU-resistant cell cancer lines or colon CA

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)
  • ELISA
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Thymidylate Synthase Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
DNA Repair
Human
IgG2a κ
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
TS 106
Unconjugated
Recombinant human TS enzyme
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions