missing translation for 'onlineSavingsMsg'
Learn More

Thymolphthalein Monophosphoric Acid Disodium Salt (>85%), TRC

Product Code. 30449208
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449208 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449208 Supplier Toronto Research Chemicals Supplier No. TRCT413078500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thymolphthalein Monophosphoric Acid Disodium Salt
CAS 123359-43-3
Molecular Formula C28H29Na2O7P •xH2O
Quantity 500 mg
Synonym 3-[4-Hydroxy-2-methyl-5-(1-methylethyl)phenyl]-3-[2-methyl-5-(1-methylethyl)-4-(phosphonooxy)phenyl]-1(3H)-isobenzofuranone Disodium Salt Hydrate
InChI Formula InChI=1S/C28H31O7P.2Na.H2O/c1-15(2)20-13-23(17(5)11-25(20)29)28(22-10-8-7-9-19(22)27(30)34-28)24-14-21(16(3)4)26(12-18(24)6)35-36(31,32)33;;;/h7-16,29H,1-6H3,(H2,31,32,33);;;1H2/q;2*+1;/p-2
SMILES O=C1OC(C2=C1C=CC=C2)(C3=CC(C(C)C)=C(O)C=C3C)C4=CC(C(C)C)=C(O[P](O[Na])(O[Na])=O)C=C4C
IUPAC Name disodium;[4-[1-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-3-oxo-2-benzofuran-1-yl]-5-methyl-2-propan-2-ylphenyl] phosphate;hydrate
Molecular Weight (g/mol) 554.481802
Formula Weight 572.1552
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.