missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroglobulin Antibody (SPM517) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34777-0.1mg
This item is not returnable.
View return policy
Description
Thyroglobulin Monoclonal antibody specifically detects Thyroglobulin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Specifications
| Thyroglobulin | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 0.5-1 μg/million cells, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready | |
| AITD3TGN, TDH3, Tg, thyroglobulin | |
| Human thyroid follicular cells | |
| 0.1 mg | |
| Cancer, Cell Biology | |
| 7038 | |
| Store at -20°C to -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| SPM517 | |
| Unconjugated | |
| 10 mM PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction