missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroglobulin Antibody (TGB04 + TGB05), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34724MFV450
This item is not returnable.
View return policy
Description
Thyroglobulin Monoclonal antibody specifically detects Thyroglobulin in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| Thyroglobulin | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| TGB04 + TGB05 | |
| Flow Cytometry | |
| AITD3TGN, TDH3, Tg, thyroglobulin | |
| Human thyroid follicular cells | |
| 0.1 mL | |
| Cancer, Cell Biology | |
| 7038 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu