missing translation for 'onlineSavingsMsg'
Learn More

Thyroxine (100% pure T4 reference standard (free of T3)), TRC

Product Code. 30134452
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134452 5 mg 5mg
30135755 25 mg 25mg
18083520 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 30134452 Supplier Toronto Research Chemicals Supplier No. TRCT4256505MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-48-9
Molecular Formula C15 H11 I4 N O4
Molecular Weight (g/mol) 776.87
Synonym Thyroxine, L- (8CI), (-)-Thyroxine, 3,3',5,5'-Tetraiodo-L-thyronine, Henning, L-T4, L-Thyroxin, L-Thyroxine, Levothyroxine, Levotiron, NSC 36397, T4, T4 (hormone), THX, Tetraiodothyronine, Thyrax, Thyreoideum, Thyroxin, Thyroxinal, Thyroxine, L-Tyrosine, O-(4-hydroxy-3,5-diiodophenyl)-3,5-diiodo-
IUPAC Name (2S)-2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoic acid
SMILES N[C@@H](Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H11I4NO4/c16-8-4-7(5-9(17)13(8)21)24-14-10(18)1-6(2-11(14)19)3-12(20)15(22)23/h1-2,4-5,12,21H,3,20H2,(H,22,23)/t12-/m0/s1
Formula Weight 776.6867
Chemical Name or Material Levothyroxine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.