Learn More
Thyroxine (100% pure T4 reference standard (free of T3)), TRC
Product Code. 30135755
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135755 25 mg 25mg
30134452 5 mg 5mg
18083520 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 30135755 Supplier Toronto Research Chemicals Supplier No. TRCT42565025MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51-48-9
Molecular Formula C15 H11 I4 N O4
Molecular Weight (g/mol) 776.87
Synonym Thyroxine, L- (8CI), (-)-Thyroxine, 3,3',5,5'-Tetraiodo-L-thyronine, Henning, L-T4, L-Thyroxin, L-Thyroxine, Levothyroxine, Levotiron, NSC 36397, T4, T4 (hormone), THX, Tetraiodothyronine, Thyrax, Thyreoideum, Thyroxin, Thyroxinal, Thyroxine, L-Tyrosine, O-(4-hydroxy-3,5-diiodophenyl)-3,5-diiodo-
IUPAC Name (2S)-2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoic acid
SMILES N[C@@H](Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1)C(=O)O
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C15H11I4NO4/c16-8-4-7(5-9(17)13(8)21)24-14-10(18)1-6(2-11(14)19)3-12(20)15(22)23/h1-2,4-5,12,21H,3,20H2,(H,22,23)/t12-/m0/s1
Formula Weight 776.6867
Chemical Name or Material Levothyroxine