missing translation for 'onlineSavingsMsg'
Learn More

Thyroxine, TRC

Product Code. 30133895
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133895 5 g 5g
18030830 500 mg 500mg
17939949 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 30133895 Supplier Toronto Research Chemicals Supplier No. TRCT4256005G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-48-9
Molecular Formula C15 H11 I4 N O4
Molecular Weight (g/mol) 776.87
Synonym Thyroxine, L- (8CI), (-)-Thyroxine, 3,3',5,5'-Tetraiodo-L-thyronine, Henning, L-T4, L-Thyroxin, L-Thyroxine, Levothyroxine, Levotiron, NSC 36397, T4, T4 (hormone), THX, Tetraiodothyronine, Thyrax, Thyreoideum, Thyroxin, Thyroxinal, Thyroxine, L-Tyrosine, O-(4-hydroxy-3,5-diiodophenyl)-3,5-diiodo-
IUPAC Name (2S)-2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoic acid
SMILES N[C@@H](Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C15H11I4NO4/c16-8-4-7(5-9(17)13(8)21)24-14-10(18)1-6(2-11(14)19)3-12(20)15(22)23/h1-2,4-5,12,21H,3,20H2,(H,22,23)/t12-/m0/s1
Formula Weight 776.6867
Percent Purity >95%
Chemical Name or Material Levothyroxine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.