missing translation for 'onlineSavingsMsg'
Learn More

Tiafenacil Metabolite M-36, TRC

Product Code. 30455055
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455055 1 mg 1mg
30449305 5 mg 5mg
30457211 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455055 Supplier Toronto Research Chemicals Supplier No. TRCT4363301MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tiafenacil Metabolite M-36
Analyte or Component Names Tiafenacil Metabolite M-36
Shipping Condition Room Temperature
Molecular Formula C15 H11 Cl F4 N2 O5 S
Synonym 2-((2-Chloro-4-fluoro-5-(3-methyl-2,6-dioxo-4-(trifluoromethyl)-3,6-dihydropyrimidin-1(2H)-yl)phenyl)sulfinyl)propanoic acid
InChI Formula InChI=1S/C15H11ClF4N2O5S/c1-6(13(24)25)28(27)10-4-9(8(17)3-7(10)16)22-12(23)5-11(15(18,19)20)21(2)14(22)26/h3-6H,1-2H3,(H,24,25)
Solution Type Neat
SMILES CC(C(=O)O)S(=O)c1cc(N2C(=O)C=C(N(C)C2=O)C(F)(F)F)c(F)cc1Cl
IUPAC Name 2-[2-chloro-4-fluoro-5-[3-methyl-2,6-dioxo-4-(trifluoromethyl)pyrimidin-1-yl]phenyl]sulfinylpropanoic acid
Industry Type Water Analysis
Molecular Weight (g/mol) 442.7699
Quantity 1 mg
Formula Weight 442.0013
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.