missing translation for 'onlineSavingsMsg'
Learn More

Bandelin Electronic™ Tickopur R 33 Universal Cleaners

Product Code. 10425261
Change view
Click to view available options
Volume (Metric):
2 L
Unit Size:
2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10425261 2 L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10425261 Supplier Bandelin Electronic™ Supplier No. 883
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Product Type Universal Cleaner
Form Liquid
pH 9.9
Product Line Tickopur
Appearance Clear Pale Yellow
Container Type Polyethylene Henkel bottle
For Use With (Application) For Cleaning Exploded and Swallowed Submitted Machinery, Carburetor, Equipment and Parts, Windows, Glasses
Quantity 1 Ea.
Volume (Metric) 2 L
compliance-icons
Product Identifier
  • TICKOPUR R 33
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.