missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGIT Antibody (741182), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7898F
This item is not returnable.
View return policy
Description
TIGIT Monoclonal antibody specifically detects TIGIT in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
Specifications
| TIGIT | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
| 741182 | |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
| T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM | |
| Mouse myeloma cell line NS0-derived recombinant human TIGIT, Met1-Pro141, Accession # Q495A1 | |
| 0.1 mL | |
| Cancer, Immunology, Virology Bacteria and Parasites | |
| 201633 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction