missing translation for 'onlineSavingsMsg'
Learn More

TIGIT Mouse, APC, Clone: 1G9, enQuireBio™

Mouse Monoclonal Antibody

183.75€ - 394.80€

Specifications

Antigen TIGIT
Clone 1G9
Concentration 0.2mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15932228
View Documents
EnquireBio
QAB57-APC-25ug
25 μg
183.75€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15922228
View Documents
EnquireBio
QAB57-APC-100ug
100 μg
394.80€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

TIGIT
0.2mg/mL
Monoclonal
Mouse
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
100043314
IgG1 κ
Protein A/G Purified
1G9
Flow Cytometry
APC
RUO
XP_017172323
Tigit
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.