missing translation for 'onlineSavingsMsg'
Learn More

Tiglic Acid, TRC

Product Code. 30449779
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449779 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449779 Supplier Toronto Research Chemicals Supplier No. TRCT4400505G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tiglic Acid
CAS 80-59-1
Molecular Formula C5 H8 O2
Quantity 5 g
Synonym 2-Butenoic acid','2-methyl-','(2E)-, 2-Butenoic acid','2-methyl-','(E)-, Crotonic acid','2-methyl-','(E)- (8CI), Tiglic acid (6CI,7CI), (2E)-2-Methyl-2-butenoic acid, (E)-2,3-Dimethylacrylic acid, (E)-2-Methyl-2-butenoic acid, (E)-2-Methylcrotonic acid, (E)-α-Methylcrotonic acid, Cevadic acid, NSC 44235, NSC 8999, Tiglinic acid, trans-2,3-Dimethylacrylic acid, trans-2-Methyl-2-butenoic acid, trans-2-Methylcrotonic acid, trans-α,β-Dimethylacrylic acid
InChI Formula InChI=1S/C5H8O2/c1-3-4(2)5(6)7/h3H,1-2H3,(H,6,7)/b4-3+
SMILES C\C=C(/C)\C(=O)O
IUPAC Name (E)-2-methylbut-2-enoic acid
Molecular Weight (g/mol) 100.12
Formula Weight 100.0524
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.