missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMM44 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
221.00€ - 470.00€
Specifications
| Antigen | TIMM44 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18656202
|
Novus Biologicals
NBP2-94065-0.02ml |
0.02 mL |
221.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18609831
|
Novus Biologicals
NBP2-94065-0.1ml |
0.1 mL |
470.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIMM44 Polyclonal antibody specifically detects TIMM44 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| TIMM44 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 10469 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| MIMT44, mitochondrial import inner membrane translocase subunit TIM44, mitochondrial inner membrane translocase, TIM44DKFZp686H05241, translocase of inner mitochondrial membrane 44 homolog (yeast) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 153-452 of human TIMM44 (NP_006342.2). AKTAKQSAESVSKGGEKLGRTAAFRALSQGVESVKKEIDDSVLGQTGPYRRPQRLRKRTEFAGDKFKEEKVFEPNEEALGVVLHKDSKWYQQWKDFKENNVVFNRFFEMKMKYDESDNAFIRASRALTDKVTDLLGGLFSKTEMSEVLTEILRVDPAFDKDRFLKQCENDIIPNVLEAMISGELDILKDWCYEATYSQLAHPIQQAKALGLQFHSRILDIDNVDLAMGKMMEQGPVLIITFQAQLVMVVRNPKGEVVEGDPDKVLRMLYVWALCRDQDELNPYAAWRLLDISASSTEQIL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title