missing translation for 'onlineSavingsMsg'
Learn More

TIMP-2 Antibody (TIMP2/2044), PE, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-08714PE

Product Code. 18650628

  • 702.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TIMP-2 Monoclonal antibody specifically detects TIMP-2 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TIMP-2
Monoclonal
PE
CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP-2, Tissue inhibitor of metalloproteinases 2
Recombinant human TIMP-2 protein fragment (aa 48-220) (exact sequence is proprietary) (Uniprot: P16035)
0.1 mL
Angiogenesis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers
7077
Store at 4°C in the dark.
IgG1 κ
ELISA, Immunohistochemistry (Paraffin), Protein Array
TIMP2/2044
PBS
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.