missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP-4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12326-1-AP-150UL
This item is not returnable.
View return policy
Description
Tissue inhibitor of metalloproteinase 4 (TIMP4) is encoded by the gene TIMP4 located on chromosome 3. It belongs to the family of TIMP genes and is known to inhibit the activity of matrix metalloproteinases. Matrix metalloproteinases (MMP) are a group of proteinases that degrade the components of extracellular matrix. TIMP4 irreversibly binds to MMPs and inactivates them with the help of zinc as a cofactor. The netrin domain of TIMP4 is involved in platelet recruitment and aggregation. Other biological functions include response to cytokine and peptide hormone stimuli and involvement with the ovulation cycle and CNS development.Specifications
| TIMP-4 | |
| Polyclonal | |
| Unconjugated | |
| TIMP4 | |
| Metalloproteinase inhibitor 4, TIMP 4, TIMP4, TIMP-4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7079 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99727 | |
| TIMP4 | |
| TIMP-4 Fusion Protein Ag2927 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction