missing translation for 'onlineSavingsMsg'
Learn More

Tinopal RBS 200, TRC

Product Code. 30457224
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457224 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457224 Supplier Toronto Research Chemicals Supplier No. TRCT44409510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tinopal RBS 200
CAS 6416-68-8
Molecular Formula C24 H16 N3 Na O3 S
Quantity 10 mg
Synonym 4-(2H-Naphtho[1,2-d]triazol-2-yl)-2-stilbenesulfonic Acid Sodium Salt, ,'5-(2H-Naphtho[1,2-d]triazol-2-yl)-2-(2-phenylethenyl)-benzenesulfonic Acid Sodium Salt, C.I. Fluorescent Brightening Agent 46','Sodium Salt, 2H-Naphtho[1,2-d]triazole','benzenesulfonic acid deriv., 2-(4-Styryl-3-sulfophenyl)-2H-naphtho[1,2-d]triazole Sodium Salt, 4-(Naphtho-1',2', 4,5-triazol-2-yl)-2-sulfostilbene Sodium Salt, C.I. 40645, C.I. Fluorescent Brightener 46, Fluorescent Brightener 46, Pylaklor White S 15A, Sodium 2-(4-styryl-3-sulfophenyl)-2H-naphtho[1,2-d]triazole, Sodium 4-(2H-Naphtho[1,2-d]triazol-2-yl)stilbene-2-sulfonate, Tinopal RBS
InChI Formula InChI=1S/C24H17N3O3S.Na/c28-31(29,30)23-16-20(14-12-19(23)11-10-17-6-2-1-3-7-17)27-25-22-15-13-18-8-4-5-9-21(18)24(22)26-27;/h1-16H,(H,28,29,30);/q;+1/p-1/b11-10+;
SMILES [Na]OS(=O)(=O)c1cc(ccc1\C=C\c2ccccc2)n3nc4ccc5ccccc5c4n3
IUPAC Name [5-benzo[e]benzotriazol-2-yl-2-[(E)-styryl]phenyl]sulfonyloxysodium
Molecular Weight (g/mol) 449.46
Formula Weight 449.081
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.