missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tioconazole, 98+%

Potent inhibitor of cytochrome-P450

Brand:  Thermo Scientific™ J60459.03

Quantity 1 g

 View more versions of this product

Product Code. 15453229

  • 120.00€ / 1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

65899-73-2
Thioconazole
Potent inhibitor of cytochrome-P450
≥98%
387.71g/mol
Powder
White to Off-white

Specifications

1 g
Powder
65899-73-2
387.71g/mol
≥98%
White to Off-white
Potent inhibitor of cytochrome-P450
C16H13Cl3N2OS
Thioconazole
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.