Learn More
Thermo Scientific™ Tioconazole, 98+%
Potent inhibitor of cytochrome-P450
120.00€ - 180.00€
Chemical Identifiers
| CAS | 65899-73-2 |
|---|---|
| Formula Weight | 387.71g/mol |
| Synonym | Thioconazole |
| Format | Powder |
| For Use With (Application) | Potent inhibitor of cytochrome-P450 |
| Description | White to Off-white |
| Assay | ≥98% |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15453229
|
Thermo Scientific™
J60459.03 |
1 g |
120.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15463229
|
Thermo Scientific™
J60459.06 |
5 g |
180.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 65899-73-2 | |
| Thioconazole | |
| Potent inhibitor of cytochrome-P450 | |
| ≥98% |
| 387.71g/mol | |
| Powder | |
| White to Off-white |
Specifications
| White to Off-white | |
| Potent inhibitor of cytochrome-P450 | |
| C16H13Cl3N2OS | |
| Thioconazole |
| Powder | |
| 65899-73-2 | |
| 387.71g/mol | |
| ≥98% |
Safety and Handling
missing translation for 'einecsNumber' : 265-973-8
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.