Learn More
WTW™ TISAB™ Buffer Solution
Product Code. 11938094
Change view
Click to view available options
For Use With (Equipment):
Fluoride Electrode
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11938094 Fluoride Electrode 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11938094 Supplier WTW™ Supplier No. 140100
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ion strength adjustment solutions have multiple tasks: TISAB for fluoride measurements does not only adjust the pH value, by also decomplexes the fluoride-containing complex compounds. Other ion strength adjustment solutions ensure an adapted pH value as well as the addition of a suitable conducting salt to correctly capture the concentration.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Fluoride Electrode
Type TISAB Buffer Solution