Learn More
Invitrogen™ Tislelizumab Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559733
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Tislelizumab (BeiGene) is a humanized monoclonal antibody that was designed to bind to the PD-1 receptor and prevent binding of PD-L1. In addition, tislelizumab was designed to minimize binding to Fc-gamma-receptors (FcyR) on macrophages. Preclinical studies found that binding to FcyR on macrophages compromises the anti-tumor activity of PD-1 antibodies through activation of antibody-dependent macrophage-mediated killing of T effector cells.
Specifications
| Tislelizumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| BGB-A317 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG4SP |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.