Learn More
R&D Systems™ Tissue Glue Kit
Product Code. 18620494 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18620494 50 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18620494 Supplier R&D Systems™ Supplier No. NBP312174
  • ONLINE EXCLUSIVE PRICE
    562.96€
    478.80€ / 1 sets
    Save 84.16€
    14% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A Novus product, part of R&D Systems.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50 mL
Applications Immunohistochemistry, Immunohistochemistry-Paraffin
Product Line Novus