Learn More
Hanna Instruments™ Waterproof Portable pH Meter
Product Code. 11733403
Change view
Click to view available options
Range (pH):
-2 to 16
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range (pH) unitSize
11733403 -2 to 16 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11733403 Supplier Hanna Instruments™ Supplier No. HI991001N
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Features a large, dual level LCD display for easy viewing of the pH, temperature, and status indicators. Hanna Instruments™ Waterproof Portable pH Meter offers a simplified two-button operation. Compact, waterproof, and portable.
Compact, waterproof, portable pH/temperature meter that is supplied complete including a rugged carrying case and a unique titanium body pH electrode that has a built-in temperature sensor.
  • Large, dual level LCD display for easy viewing of the pH, temperature, and status indicators
  • Simplified two-button operation
  • Exceptionally long battery life of 1200 hours of continuous use
Specifications
Accuracy (Temperature) ±0.5°C (up to 60°C); ±1.0°C (outside), ±1°F (up to 140°F);
Accuracy (pH) ±0.02
For Use With (Application) Automatic one or two point pH calibration with auto buffer recognition
Range (pH) -2 to 16
Resolution (pH) 0.01
Calibration Points 4.01, 7.01, 10.01 or 4.01, 6.86, 9.18
Temperature Range (Metric) -5°C to +105°C
Temperature Range (English) 23°F to 221°F
Resolution (Temperature) 0.1°C, 0.1°F
Display Type LCD
Dimensions (L x W x H) 152 x 58 x 30 mm
Model HI991001
Power Supply Yes
Probe Type Cable
Resolution 0.01pH
Detection Range -2.0 to 16.0 pH, 1.5 V, -5.5°C to +105.0°C/23.0°F to 221.0°F Temperature
Temperature Compensation Automatic from -5 to 105ºC°C
Battery Type 1.5V
Temperature Sensor Pre-amplified pH/ORP probe with internal
Battery Life 1200 hr.
Voltage 1.5 V