Learn More
MP Biomedicals™ Titertek™ Finnpipette™ Multistepper Pipet Tips
Product Code. 11489615 Shop All MP Biomedicals Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
Unit Size:
Pack of 1000
Pack of 960
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Format Unit Size
11489615 Non-sterile Bulk Pack of 1000
11409625 Sterile Rack Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11489615 Supplier MP Biomedicals™ Supplier No. 0977943C2
  • ONLINE EXCLUSIVE PRICE
    166.00€
    157.50€ / Pack of 1000
    Save 8.50€
    5% Off
Estimated Shipment: 21-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Hydrophobic tips with very low coefficient of friction
Specifically for the Titertek™/Labsystems 8-channel Multistepper pipette
  • Made from virgin polypropylene material
  • Comes with robot-compatible (large-micron) hydrophobic filters to eliminate instrument contamination
  • 96 tips per rack
  • Volume: 1500μL
Specifications
Volume (Metric) 1500 μL
Sterility Non-sterile
Color Natural
Format Bulk
Material Polypropylene