Learn More
Heidolph™ Titramax 1000 Package
Product Code. 10355582 Shop All Heidolph Instruments Products
Change view
Click to view available options
Speed Range:
150 to 1350 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range Unit Size
10355582 150 to 1350 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10355582 Supplier Heidolph™ Supplier No. 5441220900
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Includes Titramax 1000, heating module and flat incubator hood. The Titramax 1000 Package provides all the necessary components for microplate shaking and incubator system needs in one convenient package.
Package includes:
  • One vibrating platform shaker Titramax 1000
  • One heating module
  • One flat incubator hood
Titramax 1000 provides the use of 6 microtiter plates to increase your throughput by 50%. This model is suitable for the modular incubator system and is recommended for applications which require variable temperature control up to 65°C. The 1.5 mm vibration orbit performs gentle mixing with best results using small sample volumes.
  • Set and continuously adjust the variable speed on the analog control knob from 150 to 1,350 rpm
  • An analog process timer allows for unattended operation and can be set from 1 to 120 minutes. When the set time has elapsed an acoustic alarm will sound and the operation stops
  • Titramax 1000 is a medium-sized model and accepts load capacities of 5 kg
TRUSTED_SUSTAINABILITY
Specifications
Orbit 1.5 mm
Load Bearing Capacity (Metric) 5 kg
Speed Range 150 to 1350 rpm
Temperature Range (Metric) 65°C (Max.)
Wattage 31 W
Product Type Microplate Shaker
Load Bearing Capacity (English) 11 lb.
Certifications/Compliance DIN EN 60529 IP 30
Temperature Range (English) Up to 149°F
Dimensions (D x W x H) Interior 375 x 320 x 125 mm
Dimensions (LxW) Platform 290 x 258 mm
Weight (English) 14.3 lb.
Weight (Metric) 6.5 kg
Controller Type Electronic
Model Titramax 1000
Includes Titramax 1000, Heating Module, Flat Incubator Hood
Timer Range 1 to 120 min.